Anti-Hyaluronan synthase 2 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Hyaluronan synthase 2 for WB with samples derived from Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:100 to 1:50
Type: Primary
Antigen: Hyaluronan synthase 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
- Catalog No:
- 76848-394
- Antigen Symbol:
- Hyaluronan synthase 2
- Antigen Name:
- HA synthase 2
- Antigen Synonyms:
- Hyaluronic acid synthase 2|Hyaluronan synthase 2|Hyaluronate synthase 2|has2|HAS2_HUMAN
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q92819
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 67-185 of human HAS2 (NP_005319.1)
- Sequence:
- EHRKMKKSLETPIKLNKTVALCIAAYQEDPDYLRKCLQSVKRLTYPGIKVVMVIDGNSEDDLYMMDIFSEVMGRDKSATYIWKNNFHEKGPGETDESHKESSQHVTQLVLSNKSICIMQ
- Form:
- Liquid
- Molecular Weight:
- 64 kDa
- Purification:
- Affinity purification.
- Storage Buffer:
- Supplied in Phosphate buffered saline, pH 7.3, with 50% Glycerol and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C
Frequently Bought Together