Anti-Chromogranin C / SGII Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Chromogranin C / SGII for WB and ICC/IF with samples derived from Human and Rat.
- Validated Applications: WB, ICC/IF
- Recommended Dilutions: WB: 1:500 to 1:2000, ICC/IF: 1:50 to 1:20
Type: Primary
Antigen: Chromogranin C / SGII
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
- Catalog No:
- 76864-286
- Antigen Symbol:
- Chromogranin C / SGII
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Rat
- Host:
- Rabbit
- Gene ID:
- UniprotID# P13521
- Isotype:
- IgG
- Western Blot:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 30-320 of human SCG2 (NP_003460.2)
- Sequence:
- FQRNQLLQKEPDLRLENVQKFPSPEMIRALEYIENLRQQAHKEESSPDYNPYQGVSVPLQQKENGDESHLPERDSLSEEDWMRIILEALRQAENEPQSAPKENKPYALNSEKNFPMDMSDDYETQQWPERKLKHMQFPPMYEENSRDNPFKRTNEIVEEQYTPQSLATLESVFQELGKLTGPNNQKRERMDEEQKLYTDDEDDIYKANNIAYEDVVGGEDWNPVEEKIESQTQEEVRDSKENIEKNEQINDEMKRSGQLGIQEEDLRKESKDQLSDDVSKVIAYLKRLVNA
- Form:
- Liquid
- Molecular Weight:
- 65 kDa
- Purification:
- Affinity purification.
- Storage Buffer:
- Supplied in Phosphate buffered saline, pH 7.3, with 50% Glycerol and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C
- Tested Applications:
- ICC/IF
Frequently Bought Together