Anti-Zinc Alpha 2 Glycoprotein Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to Zinc Alpha 2 Glycoprotein for WB with samples derived from Human and Mouse.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:200
Type: Primary
Antigen: Zinc Alpha 2 Glycoprotein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 76860-944
- Antigen Symbol:
- Zinc Alpha 2 Glycoprotein
- Antigen Name:
- Alpha 2 glycoprotein 1 zinc
- Antigen Synonyms:
- ZA2G|Alpha 2 glycoprotein zinc|Azgp1|Zinc Alpha 2 Glycoprotein|Alpha 2 glycoprotein 1 zinc binding|Zinc-alpha-2-glycoprotein|ZA2G_HUMAN|Zinc alpha 2 glycoprotein precursor|ZAG
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# P25311
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 21-298 of human Zinc-a2-glycoprotein (ZAG/Zinc-a2-glycoprotein (ZAG/AZGP1)) (NP_001176.1)
- Sequence:
- QENQDGRYSLTYIYTGLSKHVEDVPAFQALGSLNDLQFFRYNSKDRKSQPMGLWRQVEGMEDWKQDSQLQKAREDIFMETLKDIVEYYNDSNGSHVLQGRFGCEIENNRSSGAFWKYYYDGKDYIEFNKEIPAWVPFDPAAQITKQKWEAEPVYVQRAKAYLEEECPATLRKYLKYSKNILDRQDPPSVVVTSHQAPGEKKKLKCLAYDFYPGKIDVHWTRAGEVQEPELRGDVLHNGNGTYQSWVVVAVPPQDTAPYSCHVQHSSLAQPLVVPWEAS
- Form:
- Liquid
- Molecular Weight:
- 41 kDa
- Purification:
- Affinity purification.
- Storage Buffer:
- Supplied in Phosphate buffered saline, pH 7.3, with 50% Glycerol and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C
Specifications
Frequently Bought Together