Anti-Apolipoprotein CII / ApoC-II Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to Apolipoprotein CII / ApoC-II for WB with samples derived from Human and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:100 to 1:50
Type: Primary
Antigen: Apolipoprotein CII / ApoC-II
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
Specifications
- Catalog No:
- 76850-172
- Antigen Symbol:
- Apolipoprotein CII / ApoC-II
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Rat
- Host:
- Rabbit
- Gene ID:
- UniprotID# P02655
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 1-101 of human APOC2 (NP_000474.2)
- Sequence:
- MGTRLLPALFLVLLVLGFEVQGTQQPQQDEMPSPTFLTQVKESLSSYWESAKTAAQNLYEKTYLPAVDEKLRDLYSKSTAAMSTYTGIFTDQVLSVLKGEE
- Form:
- Liquid
- Molecular Weight:
- 11 kDa
- Purification:
- Affinity purification.
- Storage Buffer:
- Supplied in Phosphate buffered saline, pH 7.3, with 50% Glycerol and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C
Specifications
Frequently Bought Together