Anti-SCF Protein Rat Monoclonal Antibody [clone: KT117] (HRP (Horseradish Peroxidase))
Catalog # 77041-936
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Symbol:SCF Protein
- Clonality:Monoclonal
- Clone:KT117
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rat
- Isotype:IgG2b
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P21583
- Storage Buffer:Supplied in Phosphate buffered saline with 0.01% Thimerosal
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant soluble human Kit ligand
- Tested Applications:Sandwich ELISA
- Purification:IgG is purified through a Protein A column and labelled with horseradish peroxidase.
- Cat. No.:77041-936
- Packaging:Plastic vial
Specifications
About this item
Rat monoclonal (KT117) antibody to Kit ligand (SCF) (HRP) for sELISA with samples derived from Human.
- Validated Applications: sELISA
- Recommended Dilutions: sELISA: 1:1000
Type: Primary
Antigen: SCF Protein
Clonality: Monoclonal
Clone: KT117
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human