Anti-TCF-4 Rabbit Polyclonal Antibody
Supplier: Antibodies.com
Certificates
About this item
Rabbit polyclonal antibody to TCF-4 for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: TCF-4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77225-892
- Antibody Type:
- Primary
Specifications
Frequently Bought Together