Anti-WIPI1 Rabbit Monoclonal Antibody [clone: ARC1652]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC1652] antibody to WIPI1 for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: WIPI1
Clonality: Monoclonal
Clone: ARC1652
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77224-988
- Antibody Type:
- Primary
Specifications
Frequently Bought Together