Anti-GTF2F2 Rabbit Monoclonal Antibody [clone: ARC2513]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC2513] antibody to GTF2F2 for WB and IHC with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC
- Recommended Dilutions: WB: 1:500 to 1:1,000, IHC: 1:50 to 1:200
Type: Primary
Antigen: GTF2F2
Clonality: Monoclonal
Clone: ARC2513
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77223-788
- Antibody Type:
- Primary
Specifications
Frequently Bought Together