Anti-Torsin A/DYT1 Rabbit Monoclonal Antibody [clone: ARC1645]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC1645] antibody to Torsin A/DYT1 for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:100 to 1:500
Type: Primary
Antigen: Torsin A / DYT1
Clonality: Monoclonal
Clone: ARC1645
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77222-738
- Antibody Type:
- Primary
Specifications
Related Information
Frequently Bought Together