Anti-TIP49A Rabbit Monoclonal Antibody [clone: ARC1247]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC1247] antibody to TIP49A for WB, IHC, IP and ChIP with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, IP, ChIP
- Recommended Dilutions: WB: 1:500 to 1:2,000, IHC: 1:50 to 1:200, ChIP: 1:50 to 1:200
Type: Primary
Antigen: TIP49A
Clonality: Monoclonal
Clone: ARC1247
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77221-578
- Antibody Type:
- Primary
Specifications
Frequently Bought Together