Anti-FGFR3 (phospho Tyr724) Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to FGFR3 (phospho Tyr724) for WB with samples derived from Human and Mouse.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: FGFR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77218-738
- Antigen Symbol:
- FGFR3
- Antigen Name:
- ACH
- Antigen Synonyms:
- CD 333|FGFR-3|CD333 antigen|CEK2|FGFR3_HUMAN|CEK 2|FGFR 3|FGFR3|CD333
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# P22607
- Isotype:
- IgG
- Western Blot:
- Yes
- Modification:
- phospho Tyr724
- Immunogen:
- A synthetic phosphorylated peptide around Y724 of human FGFR3 (NP_000133.1).
- Sequence:
- DLYMI
- Form:
- Liquid
- Molecular Weight:
- 125 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.01% Thiomersal
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C
Specifications
Frequently Bought Together