Anti-MLX-interacting protein Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to MLX-interacting protein for WB with samples derived from Human and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: MLX-interacting protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
Specifications
- Catalog No:
- 77217-954
- Antigen Symbol:
- MLX-interacting protein
- Antigen Name:
- MLX-interacting protein
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Rat
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q9HAP2
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- A synthetic peptide corresponding to a sequence within amino acids 50-150 of human MLXIP (NP_055753.3).
- Sequence:
- ARAHASAAPPPPRAGPGREEPPRRQQIIHSGHFMVSSPHREHPPKKGYDFDTVNKQTCQTYSFGKTSSCHLSIDASLTKLFECMTLAYSGKLVSPKWKNFK
- Form:
- Liquid
- Molecular Weight:
- 91 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.01% Thiomersal
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C
Specifications
Frequently Bought Together