Anti-Sodium Iodide Symporter Mouse Monoclonal Antibody [clone: 14F]
Catalog # 77172-830
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Na(+)/I(-) cotransporter
- Antigen Symbol:Sodium Iodide Symporter
- Clonality:Monoclonal
- Clone:14F
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q92911
- Antigen Synonyms:NIS|Sodium Iodide Symporter|Na+/I- cotransporter|SC5A5_HUMAN|Na(+)/I(-) symporter|Slc5a5|Na+/I-symporter|Sodium-iodide symporter|Sodium/iodide cotransporter
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 50% Glycerol and 0.09% Sodium Azide
- Molecular Weight:~97 kDa (non-glycosylated version at 68 kDa)
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Mannose binding protein hNIS fusion (AA468-643).
- Tested Applications:IHC
- Purification:Protein G purification
- Cat. No.:77172-830
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [14F] antibody to Sodium Iodide Symporter for WB, IHC and Antibody Microarray with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, Antibody Microarray
- Recommended Dilutions: WB: 1:1,000, IHC: 1:1,000
Type: Primary
Antigen: Sodium Iodide Symporter
Clonality: Monoclonal
Clone: 14F
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat