Anti-TRAP1 Mouse Monoclonal Antibody [clone: 3H4-2H6]
Catalog # 77172-790
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Heat shock protein 75 kDa
- Antigen Symbol:TRAP1
- Clonality:Monoclonal
- Clone:3H4-2H6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q12931
- Antigen Synonyms:TNFR-associated protein 1|TRAP1_HUMAN|Heat shock protein 75 kDa, mitochondrial|HSP 75|TRAP-1|TNF receptor associated protein 1|HSP75|TRAP1|HSP90L
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 50% Glycerol and 0.09% Sodium Azide
- Molecular Weight:~75 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Purified recombinant Human TRAP2.
- Tested Applications:ICC/IF
- Purification:Protein G purification
- Cat. No.:77172-790
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [3H4-2H6] antibody to TRAP1 for WB and ICC/IF with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ICC/IF
- Recommended Dilutions: WB: 1:1,000, ICC/IF: 1:100
Type: Primary
Antigen: TRAP1
Clonality: Monoclonal
Clone: 3H4-2H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Mouse;Rat