Anti-SQSTM1/p62 Rabbit Polyclonal Antibody
Catalog # 77172-770
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:A170
- Antigen Symbol:SQSTM1 / p62
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q13501
- Antigen Synonyms:EBI3 associated protein p60|EBI3-associated protein of 60 kDa|EBIAP|EBI 3 associated protein p60|FTDALS3|MGC127197|DMRV|EBI 3 associated protein of 60 kDa|EBI3 associated protein of 60 kDa
- Storage Buffer:Supplied in Phosphate Buffered Saline with 50% Glycerol and 0.09% Sodium Azide
- Molecular Weight:~60 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide from the N-terminal to mid of Human SQSTM1 (aa. 195-205).
- Tested Applications:ICC/IF
- Purification:peptide. affinity purification
- Cat. No.:77172-770
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to SQSTM1/p62 for WB and ICC/IF with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ICC/IF
- Recommended Dilutions: WB: 1:1,000, ICC/IF: 1:100
Type: Primary
Antigen: SQSTM1 / p62
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat