Order Entry
United States
ContactUsLinkComponent
Anti-ORP150 Mouse Monoclonal Antibody [clone: 6E3-2C3]
Anti-ORP150 Mouse Monoclonal Antibody [clone: 6E3-2C3]
  77172-698
undefined
Anti-ORP150 Mouse Monoclonal Antibody [clone: 6E3-2C3]
  77172-698
 :  A304973-100

 

  • Antibody Type:
    Primary
  • Antigen Name:
    150 kDa oxygen regulated protein
  • Antigen Symbol:
    ORP150
  • Clonality:
    Monoclonal
  • Clone:
    6E3-2C3
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q9Y4L1
  • Antigen Synonyms:
    glucose regulated protein 170|150 kDa oxygen-regulated protein|170 kDa glucose-regulated protein|DKFZp686N08236|FLJ94899|Grp 170|170 kDa glucose regulated protein|GRP-170|FLJ97572
  • Storage Buffer:
    Supplied in Phosphate Buffered Saline, pH 7.4, with 50% Glycerol and 0.1% Sodium Azid
  • Molecular Weight:
    ~170 kDa
  • Storage Temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping Temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant Full length Human GRP170 Protein.
  • Tested Applications:
    ICC/IF
  • Purification:
    Protein G purification
  • Cat. No.:
    77172-698
  • Packaging:
    Plastic vial

 

 

Mouse monoclonal [6E3-2C3] antibody to ORP150 for WB and ICC/IF with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, ICC/IF
  • Recommended Dilutions: WB: 1:1,000, ICC/IF: 1:100

Type: Primary
Antigen: ORP150
Clonality: Monoclonal
Clone: 6E3-2C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human;Mouse;Rat