Anti-alpha Synuclein (phospho Ser129) Rabbit Monoclonal Antibody [clone: J18]
Catalog # 77172-618
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Alpha synuclein
- Antigen Symbol:alpha Synuclein
- Clonality:Monoclonal
- Clone:J18
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P37840
- Antigen Synonyms:MGC64356|Alpha-synuclein|MGC127560|Alpha-synuclein, isoform NACP140|Non A beta component of AD amyloid|MGC110988|MGC105443|NACP|alphaSYN
- Modification:phospho Ser129
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 50% Glycerol and 0.09% Sodium Azide
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Human alpha synuclein AA 124-134: AYEMP-pS-EEGYQ-Cys.
- Tested Applications:ICC, IHC
- Purification:Affinity purification
- Cat. No.:77172-618
- Packaging:Plastic vial
Specifications
About this item
Rabbit monoclonal [J18] antibody to alpha Synuclein (phospho Ser129) for ELISA, WB, ICC and IHC with samples derived from Human and Mouse.
- Validated Applications: ELISA, WB, ICC, IHC
- Recommended Dilutions: WB: 1:500
Type: Primary
Antigen: alpha Synuclein
Clonality: Monoclonal
Clone: J18
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse