Anti-TXNDC5 Mouse Monoclonal Antibody [clone: 2E7/7]
77172-462
- Antibody Type:Primary
- Antigen Name:EndoPDI
- Antigen Symbol:TXNDC5
- Clonality:Monoclonal
- Clone:2E7/7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q8NBS9
- Antigen Synonyms:PDIA15|Endoplasmic reticulum protein ERp46|Endoplasmic reticulum resident protein 46|Protein disulfide isomerase family A member 15|STRF8|ER protein 46|Endothelial protein disulphide isomerase|MGC3178|Hcc 2
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 50% Glycerol and 0.09% Sodium Azide
- Molecular Weight:~48 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthesized peptide - 12 amino acids long (ADGEDGQDPHSK) corresponding to residues 52 - 63 of the Human EndoPDI protein.
- Tested Applications:IHC, ICC/IF
- Purification:Protein G purification
- Cat. No.:77172-462
- Packaging:Plastic vial
Mouse monoclonal [2E7/7] antibody to TXNDC5 for WB, IHC, ICC/IF, IP and ELISA with samples derived from Human.
- Validated Applications: WB, IHC, ICC/IF, IP, ELISA
- Recommended Dilutions: WB: 1:200
Type: Primary
Antigen: TXNDC5
Clonality: Monoclonal
Clone: 2E7/7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human