Anti-BVR Rabbit Polyclonal Antibody
Catalog # 77172-454
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Symbol:BVR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P53004
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 50% Glycerol and 0.09% Sodium Azide
- Molecular Weight:~40 - 42 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Human native full-length BVR.
- Tested Applications:IHC, ICC/IF
- Purification:Protein A purification
- Cat. No.:77172-454
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to BVR for WB, IHC, ICC/IF and IP with samples derived from Human.
- Validated Applications: WB, IHC, ICC/IF, IP
- Recommended Dilutions: WB: 1:2,000, ICC/IF: 1:120, IP: 1:100
Type: Primary
Antigen: BVR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human