Anti-Heme Oxygenase 1 Mouse Monoclonal Antibody [clone: 1F12-A6]
77172-294
- Antibody Type:Primary
- Antigen Name:32 kD
- Antigen Symbol:Heme Oxygenase 1
- Clonality:Monoclonal
- Clone:1F12-A6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Hamster,Pig,Rabbit,Monkey,Human,Bovine,Guinea pig,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P09601
- Antigen Synonyms:heat shock protein 32 kD|Heme Oxygenase 1|Hmox|heat shock protein 32kD|Hemox|Heme oxygenase (decycling) 1|D8Wsu38e|bK286B10|Heat shock protein
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 50% Glycerol and 0.09% Sodium Azide
- Molecular Weight:32 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Human HO-1 synthetic peptide, amino acids 1-30.
- Tested Applications:IHC, ICC/IF
- Purification:Protein G purification
- Cat. No.:77172-294
- Packaging:Plastic vial
Mouse monoclonal [1F12-A6] antibody to Heme Oxygenase 1 for WB, IHC, ICC/IF, IP and ELISA with samples derived from Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Hamster, Monkey, Porcine and Rabbit.
- Validated Applications: WB, IHC, ICC/IF, IP, ELISA
- Recommended Dilutions: WB: 1:1,000, IHC: 1:100, ICC/IF: 1:100
Type: Primary
Antigen: Heme Oxygenase 1
Clonality: Monoclonal
Clone: 1F12-A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Mouse;Rat;Bovine;Dog;Guinea pig;Hamster;Monkey;Pig;Rabbit