Anti-Myosin Light Chain 2 (phospho Ser19) Rabbit Recombinant Antibody [clone: RM484]
Catalog # 76833-950
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Cardiac myosin light chain-2
- Antigen Symbol:Myosin Light Chain 2
- Clonality:Recombinant
- Clone:RM484
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:MYL2|CMH10|MLC 2v|MYL 2|MLC2|MLRV_HUMAN|MLC-2|MLC-2v|Cardiac ventricular myosin light chain 2
- Modification:phospho Ser19
- Storage Buffer:Supplied in Phosphate Buffered Saline with 50% Glycerol, 1% BSA, and 0.09% Sodium Azide
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic phospho-peptide corresponding to human Myosin Light Chain 2 (phospho Ser19).
- Purification:Protein A chromatography
- Cat. No.:76833-950
- Packaging:Plastic vial
Specifications
About this item
Rabbit monoclonal [RM484] antibody to Myosin Light Chain 2 (phospho Ser19) for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:1,000 to 1:2,000
Type: Primary
Antigen: Myosin Light Chain 2
Clonality: Recombinant
Clone: RM484
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat