Anti-Horse IgG Goat Polyclonal Antibody
Catalog # 76836-156
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Secondary
- Antigen Symbol:Horse IgG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Horse
- Western Blot:Yes
- Size:1 mg
- Form:Liquid
- Antibody Fragment:Whole Molecule
- Storage Buffer:Supplied in Phosphate Buffered Saline with 0.09% Sodium Azide
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Tested Applications:IHC, ICC
- Purification:Antigen affinity purification
- Cat. No.:76836-156
- Packaging:Plastic vial
Specifications
About this item
Goat anti-horse IgG secondary antibody for WB, IHC, ICC and ELISA with samples derived from Horse.
- Validated Applications: WB, IHC, ICC, ELISA
- Recommended Dilutions: ELISA: 1:1,000 to 1:30,000; ELISA (capture antibody): 1:100 to 1:500, ICC: 1:200 to 1:2,000, IHC: 1:200 to 1:2,000, WB: 1:1,000 to 1:30,000
Type: Secondary
Antigen: Horse IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Horse