Anti-Goat IgG H&L Donkey Polyclonal Antibody (Biotin)
76833-624
- Antibody Type:Secondary
- Antigen Symbol:Goat IgG H&L
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Donkey
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Goat
- Western Blot:Yes
- Size:1 mg
- Form:Liquid
- Antibody Fragment:Whole Molecule
- Storage Buffer:Supplied in Phosphate Buffered Saline with 0.09% Sodium Azide
- Storage Temperature:Shipped at 4 °C. Store at 4 °C. Do not freeze.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Tested Applications:IHC, ICC
- Purification:Antigen affinity purification
- Cat. No.:76833-624
- Packaging:Plastic vial
Donkey anti-goat IgG H&L (Biotin) secondary antibody for WB, IHC, ICC and ELISA with samples derived from Goat.
- Validated Applications: WB, IHC, ICC, ELISA
- Recommended Dilutions: ELISA: 1:20,000 to 1:400,000, ICC: 1:200 to 1:1,000, IHC: 1:500 to 1:5,000, WB: 1:20,000 to 1:400,000
Type: Secondary
Antigen: Goat IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Goat