To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Secondary
- Antigen Symbol:Sheep IgG H&L
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Sheep
- Size:1 mg
- Environmentally Preferable:
- Form:Liquid
- Antibody Fragment:Whole Molecule
- Storage Buffer:Supplied in Phosphate Buffered Saline with 0.09% Sodium Azide
- Storage Temperature:Store in the dark at 4 °C. Avoid prolonged exposure to light. Do not freeze.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Tested Applications:IHC, ICC
- Purification:Antigen affinity purification
- Cat. No.:76834-098
- Packaging:Plastic vial
Specifications
About this item
Rabbit anti-sheep IgG H&L (FITC) secondary antibody for IHC, ICC, Flow Cytometry and IF with samples derived from Sheep.
- Validated Applications: IHC, ICC, Flow Cytometry, IF
- Recommended Dilutions: Flow Cytometry: 1:50 to 1:200, ICC: 1:50 to 1:500, IF: 1:50 to 1:500, IHC: 1:50 to 1:500
Type: Secondary
Antigen: Sheep IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Sheep