Anti-Rabbit IgG H&L Donkey Polyclonal Antibody
76836-402
- Antibody Type:Secondary
- Antigen Symbol:Rabbit IgG H&L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Donkey
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rabbit
- Western Blot:Yes
- Size:500 µg
- Form:Liquid
- Antibody Fragment:F(ab')2
- Storage Buffer:Supplied in Phosphate Buffered Saline with 0.09% Sodium Azide
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Tested Applications:IHC, ICC
- Purification:Antigen affinity purification
- Cat. No.:76836-402
- Packaging:Plastic vial
Donkey F(ab')2 anti-rabbit IgG H&L secondary antibody for WB, IHC, ICC and ELISA with samples derived from Rabbit.
- Validated Applications: WB, IHC, ICC, ELISA
- Recommended Dilutions: ELISA: 1:1,000 to 1:30,000; ELISA (capture antibody): 1:100 to 1:500, ICC: 1:200 to 1:2,000, IHC: 1:200 to 1:2,000, WB: 1:1,000 to 1:30,000
Type: Secondary
Antigen: Rabbit IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Rabbit