To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Secondary
- Antigen Symbol:Rabbit IgG F(ab')2
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rabbit
- Western Blot:Yes
- Size:500 µg
- Environmentally Preferable:
- Form:Liquid
- Antibody Fragment:F(ab')2
- Storage Buffer:Supplied in Phosphate Buffered Saline with 0.2% BSA and 0.09% Sodium Azide
- Storage Temperature:Shipped at 4 °C. Store at 4 °C. Do not freeze.
- Concentration:0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Tested Applications:IHC, ICC
- Purification:Antigen affinity purification
- Cat. No.:76836-366
- Packaging:Plastic vial
Specifications
About this item
Goat F(ab')2 anti-rabbit IgG F(ab')2 (Biotin) secondary antibody for WB, IHC, ICC and ELISA with samples derived from Rabbit.
- Validated Applications: WB, IHC, ICC, ELISA
- Recommended Dilutions: ELISA: 1:20,000 to 1:400,000, ICC: 1:200 to 1:1,000, IHC: 1:500 to 1:5,000, WB: 1:20,000 to 1:400,000
Type: Secondary
Antigen: Rabbit IgG F(ab')2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit