Anti-Rabbit IgG H&L Donkey Polyclonal Antibody
Catalog # 76836-108
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Secondary
- Antigen Symbol:Rabbit IgG H&L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Donkey
- Isotype:IgG
- Reactivity:Rabbit
- Size:2 ml
- Form:Liquid
- Antibody Fragment:Whole Molecule
- Storage Buffer:Supplied as an aliquot of serum with 0.09% Sodium Azide
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Tested Applications:IEP, Double Immudiffusion
- Cat. No.:76836-108
- Packaging:Plastic vial
Specifications
About this item
Donkey anti-rabbit IgG H&L secondary antibody for IEP and Double Immunodiffusion with samples derived from Rabbit.
- Validated Applications: IEP, Double Immunodiffusion
- Recommended Dilutions: DD: Neat, IEP: Neat
Type: Secondary
Antigen: Rabbit IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Rabbit