To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:Pan Trk
- Clonality:Recombinant
- Clone:RM423
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P04629
- Storage Buffer:Supplied in Phosphate Buffered Saline with 50% Glycerol, 1% BSA, and 0.09% Sodium Azide
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to the C terminus of human TRK.
- Tested Applications:IHC
- Purification:Protein A chromatography
- Cat. No.:76835-132
- Packaging:Plastic vial
Recombinant rabbit monoclonal [RM423] antibody to Pan Trk for IHC and WB with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC, WB
- Recommended Dilutions: IHC: 1:100 to 1:200, WB: 1:1,000 to 1:2,000
Type: Primary
Antigen: Pan Trk
Clonality: Recombinant
Clone: RM423
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat