Anti-Musashi 2 Rabbit Recombinant Antibody [clone: RM422]
76834-276
- Antibody Type:Primary
- Antigen Name:FLJ36569
- Antigen Symbol:Musashi 2
- Clonality:Recombinant
- Clone:RM422
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:UniprotID# Q96DH6
- Antigen Synonyms:Musashi, Drosophila, homolog of, 2|Musashi RNA binding protein 2|MGC3245|Musashi homolog 2|MSI2/HOXA9 fusion gene, included|MSI2|MSI2H_HUMAN|MSI2H|Musashi 2
- Storage Buffer:Supplied in Phosphate Buffered Saline with 50% Glycerol, 1% BSA, and 0.09% Sodium Azide
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to the internal region of human Musashi 2.
- Tested Applications:IHC
- Purification:Protein A chromatography
- Cat. No.:76834-276
- Packaging:Plastic vial
Recombinant rabbit monoclonal [RM422] antibody to Musashi 2 for IHC and WB with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC, WB
- Recommended Dilutions: IHC: 1:100 to 1:400, WB: 1:1,000 to 1:2,000
Type: Primary
Antigen: Musashi 2
Clonality: Recombinant
Clone: RM422
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat