Anti-PRDM16 Goat Polyclonal Antibody
Catalog # 76961-726
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:CMD1LL
- Antigen Symbol:PRDM16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- ChIP:Yes
- Form:Liquid
- Gene ID:63976
- Antigen Synonyms:PR domain containing protein 16|MDS1/EVI1-like gene 1|PFM13|LVNC8|MDS1/EVI1 like gene 1|PFM 13|PR domain containing 16|KIAA1675|MEL1
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.3, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-TSESGAFHPINHL
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to the C terminus of human PRDM16.
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:76961-726
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to PRDM16 for ELISA, Flow Cytometry and ChIP with samples derived from Human and Mouse.
- Validated Applications: ELISA, Flow Cytometry, ChIP
Type: Primary
Antigen: PRDM16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse