Order Entry
United States
ContactUsLinkComponent
Anti-IL-1 beta Rabbit Recombinant Antibody [clone: IL1B/7049R]
Anti-IL-1 beta Rabbit Recombinant Antibody [clone: IL1B/7049R]
Catalog # 76951-104
Supplier:  ANTIBODIES.COM LLC
Anti-IL-1 beta Rabbit Recombinant Antibody [clone: IL1B/7049R]
Catalog # 76951-104
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A278592-100

Specifications

  • Cat. No.:
    76951-104

Specifications

About this item

Recombinant rabbit monoclonal [IL1B/7049R] antibody to IL-1 beta for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: IL-1 beta
Clonality: Recombinant
Clone: IL1B/7049R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human