Anti-SP100 Mouse Monoclonal Antibody [clone: PCRP-SP100-1B9]
Catalog # 76960-658
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:DKFZp686E07254
- Antigen Symbol:SP100
- Clonality:Monoclonal
- Clone:PCRP-SP100-1B9
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P23497
- Antigen Synonyms:Nuclear autoantigen Sp100|Lysp100b|FLJ00340|Nuclear autoantigen Sp-100|FLJ34579|Nuclear autoantigen Sp 100|Nuclear dot-associated Sp100 protein|Nuclear dot associated Sp100 protein|Nuclear antigen Sp100
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:100.4 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human SP100 protein.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76960-658
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PCRP-SP100-1B9] antibody to SP100 for ELISA, WB, IF, IP, Flow Cytometry and IHC-P with samples derived from Human.
- Validated Applications: ELISA, WB, IF, IP, Flow Cytometry, IHC-P
- Recommended Dilutions: WB: 1 to 2 µg/ml, IF: 0.5 to 2 µg/ml, IP: 1 to 2µg / 100 to 500µg proteins, FC: 0.5 to 2 µg/million cells, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: SP100
Clonality: Monoclonal
Clone: PCRP-SP100-1B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human