Anti-Phosphoglucomutase 5 Mouse Monoclonal Antibody [clone: PGM5/3552]
Catalog # 76962-496
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Phosphoglucomutase 5
- Antigen Symbol:Phosphoglucomutase 5
- Clonality:Monoclonal
- Clone:PGM5/3552
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Rabbit,Human,Bovine,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q15124
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:56 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human PGM5 protein.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76962-496
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PGM5/3552] antibody to Phosphoglucomutase 5 for ELISA, IF, WB and IHC-P with samples derived from Human, Bovine, Mouse, Rat and Rabbit.
- Validated Applications: ELISA, IF, WB, IHC-P
- Recommended Dilutions: IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Phosphoglucomutase 5
Clonality: Monoclonal
Clone: PGM5/3552
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human;Bovine;Mouse;Rat;Rabbit