Anti-JAZF1 Mouse Monoclonal Antibody [clone: PCRP-JAZF1-1C2]
Catalog # 76961-478
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:JAZF zinc finger 1
- Antigen Symbol:JAZF1
- Clonality:Monoclonal
- Clone:PCRP-JAZF1-1C2
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q86VZ6
- Antigen Synonyms:Zinc finger protein 802|JAZF1_HUMAN|JAZF1|Juxtaposed with another zinc finger protein 1|TIP27|ZNF802|TAK1-interacting protein 27|TAK1 interacting protein 27
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:27.08 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 1-130, of human JAZF1 protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76961-478
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PCRP-JAZF1-1C2] antibody to JAZF1 for IP, Flow Cytometry, IF, WB and IHC-P with samples derived from Human.
- Validated Applications: IP, Flow Cytometry, IF, WB, IHC-P
- Recommended Dilutions: IP: 1 to 2µg / 100 to 500µg proteins, Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: JAZF1
Clonality: Monoclonal
Clone: PCRP-JAZF1-1C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human