Anti-DAXX Mouse Monoclonal Antibody [clone: PCRP-DAXX-6A8]
Catalog # 76960-934
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:BING 2
- Antigen Symbol:DAXX
- Clonality:Monoclonal
- Clone:PCRP-DAXX-6A8
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2c
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9UER7
- Antigen Synonyms:DAP 6|BING2|Death associated protein 6|Death domain associated protein|DAXX_HUMAN|CENP-C binding protein|DAP6|Daxx
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:81.37 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human DAXX protein.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76960-934
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PCRP-DAXX-6A8] antibody to DAXX for Flow Cytometry, IF and IHC-P with samples derived from Human.
- Validated Applications: Flow Cytometry, IF, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: DAXX
Clonality: Monoclonal
Clone: PCRP-DAXX-6A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2c
Reactivity: Human