Anti-MEF2B Mouse Monoclonal Antibody [clone: PCRP-MEF2B-2F9]
76957-664
- Antibody Type:Primary
- Antigen Name:MADS box transcription enhancer factor 2
- Antigen Symbol:MEF2B
- Clonality:Monoclonal
- Clone:PCRP-MEF2B-2F9
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q02080
- Antigen Synonyms:MEF2B_HUMAN|RSRFR2|Serum response factor like protein 2|Myocyte enhancer factor 2B|Myocyte-specific enhancer factor 2B|Myocyte specific enhancer factor 2B|MEF2B|MDS1 EVI1|PRDM3
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:160 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human MEF2B protein.
- Purification:Protein A/G chromatography
- Cat. No.:76957-664
- Packaging:Plastic vial
Mouse monoclonal [PCRP-MEF2B-2F9] antibody to MEF2B for IP, WB, Flow Cytometry and IF with samples derived from Human.
- Validated Applications: IP, WB, Flow Cytometry, IF
- Recommended Dilutions: IP: 1 to 2µg / 100 to 500µg proteins, WB: 1 to 2 µg/ml, Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: MEF2B
Clonality: Monoclonal
Clone: PCRP-MEF2B-2F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human