Anti-Decorin Rabbit Recombinant Antibody [clone: DCN/7031R]
Catalog # 76958-466
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Cat. No.:76958-466
Specifications
About this item
Recombinant rabbit monoclonal [DCN/7031R] antibody to Decorin for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Decorin
Clonality: Recombinant
Clone: DCN/7031R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human