Order Entry
United States
ContactUsLinkComponent
Anti-FOXP1 Mouse Recombinant Antibody [clone: rFOXP1/6902]
Anti-FOXP1 Mouse Recombinant Antibody [clone: rFOXP1/6902]
Catalog # 76958-664
Supplier:  ANTIBODIES.COM LLC
Anti-FOXP1 Mouse Recombinant Antibody [clone: rFOXP1/6902]
Catalog # 76958-664
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A277889-100

Specifications

  • Cat. No.:
    76958-664

Specifications

About this item

Recombinant mouse monoclonal [rFOXP1/6902] antibody to FOXP1 for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: FOXP1
Clonality: Recombinant
Clone: rFOXP1/6902
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human