Anti-MED21 Mouse Monoclonal Antibody [clone: PCRP-MED21-4B5]
Catalog # 76949-990
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:hSrb7
- Antigen Symbol:MED21
- Clonality:Monoclonal
- Clone:PCRP-MED21-4B5
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q13503
- Antigen Synonyms:SRB7 suppressor of RNA polymerase B homolog|MED21_HUMAN|RNAPII complex component SRB7|MED21|Mediator of RNA polymerase II transcription subunit 21|Surb7|RNA polymerase II holoenzyme component SRB7|Mediator complex subunit 21
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:15.5 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human MED21 protein.
- Purification:Protein A/G chromatography
- Cat. No.:76949-990
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PCRP-MED21-4B5] antibody to MED21 for ELISA, IP, Flow Cytometry, IF and WB with samples derived from Human.
- Validated Applications: ELISA, IP, Flow Cytometry, IF, WB
- Recommended Dilutions: IP: 1 to 2µg / 100 to 500µg proteins, Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml
Type: Primary
Antigen: MED21
Clonality: Monoclonal
Clone: PCRP-MED21-4B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human