Anti-LY75 Mouse Monoclonal Antibody [clone: CD205/3720]
76968-776
- Antibody Type:Primary
- Antigen Name:C-type lectin domain family 13 member B
- Antigen Symbol:LY75
- Clonality:Monoclonal
- Clone:CD205/3720
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O60449
- Antigen Synonyms:gp200|CLEC 13B|CD205|CD 205|DEC205|DEC 205|DEC-205|CD205 antigen|CLEC13B
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:198 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 1,006-1,130, of human LY75 protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76968-776
- Packaging:Plastic vial
Mouse monoclonal [CD205/3720] antibody to LY75 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: LY75
Clonality: Monoclonal
Clone: CD205/3720
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human