Anti-ETS2 Mouse Monoclonal Antibody [clone: PCRP-ETS2-1D9]
76955-012
- Cat. No.:76955-012
Mouse monoclonal [PCRP-ETS2-1D9] antibody to ETS2 for ELISA, Flow Cytometry, IF and IP with samples derived from Human.
- Validated Applications: ELISA, Flow Cytometry, IF, IP
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IP: 1 to 2µg / 100 to 500µg proteins
Type: Primary
Antigen: ETS2
Clonality: Monoclonal
Clone: PCRP-ETS2-1D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human