Order Entry
United States
ContactUsLinkComponent
Anti-ATF2 Mouse Monoclonal Antibody [clone: PCRP-ATF2-1B4]
Anti-ATF2 Mouse Monoclonal Antibody [clone: PCRP-ATF2-1B4]
Catalog # 76953-092
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-ATF2 Mouse Monoclonal Antibody [clone: PCRP-ATF2-1B4]
Catalog # 76953-092
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A277565-100

Specifications

  • Cat. No.:
    76953-092

Specifications

About this item

Mouse monoclonal [PCRP-ATF2-1B4] antibody to ATF2 for Flow Cytometry, IF, IP and WB with samples derived from Human.

  • Validated Applications: Flow Cytometry, IF, IP, WB
  • Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IP: 1 to 2µg / 100 to 500µg proteins, WB: 1 to 2 µg/ml

Type: Primary
Antigen: ATF2
Clonality: Monoclonal
Clone: PCRP-ATF2-1B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human