Anti-alpha 1 Antichymotrypsin Mouse Monoclonal Antibody [clone: SERPINA3/4184]
Catalog # 76954-176
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Cat. No.:76954-176
Specifications
About this item
Mouse monoclonal [SERPINA3/4184] antibody to alpha 1 Antichymotrypsin for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: alpha 1 Antichymotrypsin
Clonality: Monoclonal
Clone: SERPINA3/4184
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human