Anti-Ankyrin 3 Mouse Monoclonal Antibody [clone: 2A8]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Mouse monoclonal (2A8) antibody to Ankyrin 3 for WB, ICC/IF and IHC with samples derived from Human and Rat.
- Validated Applications: WB, ICC/IF, IHC
- Recommended Dilutions: WB: 1:1,000 to 1:2,000, ICC/IF: 1:1,000, IHC: 1:1,000
Type: Primary
Antigen: Ankyrin 3
Clonality: Monoclonal
Clone: 2A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Rat
Specifications
- Catalog No:
- 77070-250
- 77061-868
- Antigen Symbol:
- Ankyrin 3
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 2A8
- Reactivity:
- Human, Rat
- Host:
- Mouse
- Gene ID:
- UniprotID# Q12955
- Isotype:
- IgG1
- Western Blot:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant fragment corresponding to the C-terminal 398 amino acids of human Ankyrin 3 isotype 1 (NP_066267.2), expressed in and purified from E. coli. This segment is expressed by all three Ankyrin 3 isotypes and contains the DEATH domain sequence.
- Form:
- Liquid
- Molecular Weight:
- 480 kDa, 270 kDa, 190 kDa
- Purification:
- Immunogen affinity purification
- Concentration:
- 1 mg/ml
- Storage Buffer:
- Supplied in Phosphate Buffered Saline with 50% Glycerol and 5 mM Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C
- Tested Applications:
- ICC/IF, IHC
Specifications
Frequently Bought Together