Anti-CD52 Mouse Monoclonal Antibody [clone: 4C8]
Catalog # 76959-686
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Cat. No.:76959-686
Specifications
About this item
Mouse monoclonal [4C8] antibody to CD52 for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: 1 to 4 µg/ml
Type: Primary
Antigen: CD52
Clonality: Monoclonal
Clone: 4C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human