Anti-CD15 Mouse Monoclonal Antibody [clone: FR4A5]
76967-328
- Antibody Type:Primary
- Antigen Name:3-FAL
- Antigen Symbol:CD15
- Clonality:Monoclonal
- Clone:FR4A5
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgM kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P22083, P51993
- Antigen Synonyms:CD15|FUC-TIV|Alpha-3-fucosyltransferase|ELAM 1 ligand|ELFT|fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific)|ELAM-1 ligand fucosyltransferase|FCT3A|ELAM ligand fucosyltransferase
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Myelomonocytic leukemia cells.
- Cat. No.:76967-328
- Packaging:Plastic vial
Mouse monoclonal [FR4A5] antibody to CD15 for Flow Cytometry and IF with samples derived from Human.
- Validated Applications: Flow Cytometry, IF
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: CD15
Clonality: Monoclonal
Clone: FR4A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human