Anti-Melanoma Marker (MART-1 + gp100) Mouse Monoclonal Antibody [clone: DT101 + BC199 + HMB45]
Catalog # 76968-388
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Symbol:Melanoma Marker (MART-1 + gp100)
- Clonality:Monoclonal
- Clone:DT101 + BC199 + HMB45
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:Multiple
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q16655, P40967
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:20 - 22 kDa (doublet) (MART-1): 90 - 100 kDa (gp100)
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Clone DT101 & BC199: Recombinant human MART-1 protein. Clone HMB45: Extract of pigmented melanoma metastases from lymph nodes.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76968-388
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [DT101 + BC199 + HMB45] antibody to Melanoma Marker (MART-1 + gp100) for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Melanoma Marker (MART-1 + gp100)
Clonality: Monoclonal
Clone: DT101 + BC199 + HMB45
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Multiple
Reactivity: Human