Anti-Melanoma Associated Antigen Mouse Monoclonal Antibody [clone: MAA/1414]
Catalog # 76961-528
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:FLJ10997
- Antigen Symbol:Melanoma Associated Antigen
- Clonality:Monoclonal
- Clone:MAA/1414
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:ZNF654|FLJ21142|OTTHUMP00000214069|zfp654|melanoma associated antigen|Melanoma-associated antigen|Hypothetical protein FLJ10997|ZN654_HUMAN|Zinc finger protein 654
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Proteins extracted from a melanoma.
- Purification:Protein A/G chromatography
- Cat. No.:76961-528
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [MAA/1414] antibody to Melanoma Associated Antigen for ELISA with samples derived from Human.
- Validated Applications: ELISA
- Recommended Dilutions: ELISA: 1 to 5 µg/ml
Type: Primary
Antigen: Melanoma Associated Antigen
Clonality: Monoclonal
Clone: MAA/1414
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human