Anti-Cyclin E Rabbit Recombinant Antibody [clone: CCNE1/4935R]
76952-180
- Antibody Type:Primary
- Cat. No.:76952-180
Recombinant rabbit monoclonal [CCNE1/4935R] antibody to Cyclin E for WB and IHC-P with samples derived from Human.
- Validated Applications: WB, IHC-P
- Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cyclin E
Clonality: Recombinant
Clone: CCNE1/4935R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human