Anti-WTAP Mouse Monoclonal Antibody [clone: PCRP-BATF3-1E5]
Catalog # 77181-270
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:DKFZp686F20131
- Antigen Symbol:WTAP
- Clonality:Monoclonal
- Clone:PCRP-BATF3-1E5
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q15007
- Antigen Synonyms:OTTHUMP00000017523|FL2D_HUMAN|hFL(2)D|Female-lethal(2)D homolog|PNAS 132|OTTHUMP00000017522|MGC3925|KIAA0105|Mum2
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:14.47 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-123) of human BATF3 protein (N-terminus).
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77181-270
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PCRP-BATF3-1E5] antibody to WTAP for Flow Cytometry and IHC-P with samples derived from Human.
- Validated Applications: Flow Cytometry, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: WTAP
Clonality: Monoclonal
Clone: PCRP-BATF3-1E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human